missing translation for 'onlineSavingsMsg'
Learn More

Pidolic Acid, Mikromol™

Product Code. 30051997
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051997 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051997 Supplier Mikromol Supplier No. MM3413.01
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pidolic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pidolic acid
CAS 98-79-3
Analyte or Component Names Pidolic acid
Shipping Condition Room Temperature
Molecular Formula C5 H7 N O3
Certifications/Compliance ISO 9001
Synonym 5-Oxo-L-proline, Proline, 5-oxo-, L- (8CI), (-)-2-Pyrrolidone-5-carboxylic acid, (-)-Pyroglutamic acid, (2S)-5-Oxopyrrolidine-2-carboxylic acid, (5S)-2-Oxopyrrolidine-5-carboxylic acid, (S)-(-)-2-Pyrrolidone-5-carboxylic acid, (S)-2-Pyrrolidone-5-carboxylic acid, (S)-5-Oxo-1H-pyrrolidine-2-carboxylic acid, (S)-5-Oxo-2-pyrrolidinecarboxylic acid, (S)-5-Oxoproline, (S)-Pyroglutamic acid, 2-L-Pyrrolidone-5-carboxylic acid, 2-Oxopyrrolidine-5(S)-carboxylic acid, 2-Pyrrolidinone-5-carboxylic acid, 5-Carboxy-2-pyrrolidinone, 5-Oxoproline, 5-Pyrrolidinone-2-carboxylic acid, Ajidew A 100, Glutimic acid, Glutiminic acid, L-(-)-Pyroglutamic acid, L-2-Pyrrolidone-5-carboxylic acid, L-5-Carboxy-2-pyrrolidinone, L-5-Oxo-2-pyrrolidinecarboxylic acid, L-5-Oxoproline, L-Glutimic acid, L-Glutiminic acid, L-Pyroglutamic acid, L-Pyrrolidinonecarboxylic acid, L-Pyrrolidonecarboxylic acid, NSC 143034, Oxoproline, Oxopyrrolidinecarboxylic acid, PCA, PCA 50, Pidolic acid, Pidolidone, Pyroglutamic acid, Pyrrolidinonecarboxylic acid, Pyrrolidone-5-carboxylic acid, Pyrrolidonecarboxylic acid, (S)-(-)-gamma-Butyrolactam-gamma-carboxylic acid, L-Glutamic acid, gamma-lactam
InChI Formula InChI=1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1
SMILES OC(=O)[C@@H]1CCC(=O)N1
IUPAC Name (2S)-5-oxopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 129.11
Quantity 250 mg
Formula Weight 129.0426 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.