missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pilocarpine Hydrochloride, Mikromol™
Description
Pilocarpine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Pilocarpine Hydrochloride |
| CAS | 54-71-7 |
| Analyte or Component Names | Pilocarpine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H16 N2 O2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Pilocarpine Hydrochloride, (3S,4R)-3-Ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-2(3H)-furanone monohydrochloride, (3S-cis)-3-Ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-2(3H)-furanone monohydrochloride, Pilocarpine monohydrochloride, (+)-Pilocarpine hydrochloride, Adsorbocarpine, Almocarpine, Isopto Carpine, NSC 5746, Pilocar SMP, Pilocarpal, Pilocarpine chloride, Pilocarpine hydrochloride, Pilocarpine muriate, Pilocarpinium chloride, Pilocel, Pilovisc, Salagen, Vistacarpin N |
| InChI Formula | InChI=1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10-;/m0./s1 |
| SMILES | Cl.CC[C@H]1[C@@H](Cc2cncn2C)COC1=O |
| IUPAC Name | (3S,4R)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?