missing translation for 'onlineSavingsMsg'
Learn More

PIMT Antibody [DyLight 594], Novus Biologicals Biologicals™

Product Code. 30514009 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30514009 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30514009 Supplier Novus Biologicals Supplier No. NBP338476DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIMT Polyclonal antibody specifically detects PIMT in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIMT
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Cap-specific guanine-N2 methyltransferase, CLL-associated antigen KW-2, DKFZp762A163, EC 2.1.1, EC 2.1.1.-, HCA137, Hepatocellular carcinoma-associated antigen 137, NCOA6IPSEREX-defined, nuclear receptor coactivator 6 interacting protein, Nuclear receptor coactivator 6-interacting protein, PIMTFLJ22995, PIPMT, PRIP-interacting protein PIPMT, PRIP-interacting protein with methyltransferase domain, PRIP-interacting protein with methyltransferase motif, trimethylguanosine synthase, trimethylguanosine synthase 1, trimethylguanosine synthase homolog, trimethylguanosine synthase homolog (S. cerevisiae)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 714-853 of human PIMT (NP_079107.6).,, Sequence:, RVIAIDIDPVKIALARNNAEVYGIADKIEFICGDFLLLASFLKADVVFLSPPWGGPDYATAETFDIRTMMSPDGFEIFRLSKKITNNIVYFLPRNADIDQVASLAGPGGQVEIEQNFLNNKLKTITAYFGDLIRRPASET
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 96764
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.