missing translation for 'onlineSavingsMsg'
Learn More

PIP Antibody (DBM15.52), Novus Biologicals™

Product Code. 30532798 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
1 mL
Unit Size:
0.1mL
0.50mL
1mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532798 0.1 mL 0.1mL
30532800 1 mL 1mL
30532799 0.5 mL 0.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30532798 Supplier Novus Biologicals Supplier No. NBP3451040.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIP Monoclonal antibody specifically detects PIP in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DBM15.52
Conjugate Unconjugated
Formulation PBS (pH 7.4), Tween-20, 0.2% BSA
Gene Alias GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein
Host Species Mouse
Immunogen Recombinant human GCDFP-15 protein fragment (aa 41-146).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5304
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.