missing translation for 'onlineSavingsMsg'
Learn More

PIP5K2B Antibody [DyLight 594], Novus Biologicals Biologicals™

Product Code. 30514639 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30514639 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30514639 Supplier Novus Biologicals Supplier No. NBP338497DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIP5K2B Polyclonal antibody specifically detects PIP5K2B in Human samples. It is validated for ELISA,Western Blot,Immunofluorescence
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen PIP5K2B
Applications ELISA, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias 1-phosphatidylinositol-4-phosphate kinase, 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta, Diphosphoinositide kinase 2-beta, EC 2.7.1, EC 2.7.1.149, phosphatidylinositol-4-phosphate 5-kinase, type II, beta, Phosphatidylinositol-5-phosphate 4-kinase type II beta, phosphatidylinositol-5-phosphate 4-kinase type-2 beta, phosphatidylinositol-5-phosphate 4-kinase, type II, beta, PI(5)P 4-kinase type II beta, PI5P4KB, PIP4KII-beta, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PTDINS(4)P-5-kinase, PtdIns(5)P-4-kinase isoform 2-beta
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human PIP5K2B (NP_003550.1).,, Sequence:, MSSNCTSTTAVAVAPLSASKTKTKKKHFVCQKVKLFRASEPILSVLMWGVNHTINELSNVPVPVMLMPDDFKAYSKIKVDNHLFNKENLPSRFKFKEYCPMVFRNLRERFGIDDQDYQNSVTRSAPINSDSQGRCGTRFLTTYDRRFVIKTVSSEDVAEMHNILKKYHQFIVECHGNTLLPQFLGMYRLTVDGVETYMVVTRNVFSHRLTVHRKYDLKGSTVAREASDKEKAKDLPTFKDNDFLNEGQKLHVGEESKKNFLEKLKRDVEF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8396
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.