missing translation for 'onlineSavingsMsg'
Learn More

PIP5K2B Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™

Product Code. 30514648 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30514648 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30514648 Supplier Novus Biologicals Supplier No. NBP338497JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIP5K2B Polyclonal antibody specifically detects PIP5K2B in Human samples. It is validated for ELISA,Western Blot,Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP5K2B
Applications ELISA, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias 1-phosphatidylinositol-4-phosphate kinase, 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta, Diphosphoinositide kinase 2-beta, EC 2.7.1, EC 2.7.1.149, phosphatidylinositol-4-phosphate 5-kinase, type II, beta, Phosphatidylinositol-5-phosphate 4-kinase type II beta, phosphatidylinositol-5-phosphate 4-kinase type-2 beta, phosphatidylinositol-5-phosphate 4-kinase, type II, beta, PI(5)P 4-kinase type II beta, PI5P4KB, PIP4KII-beta, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PTDINS(4)P-5-kinase, PtdIns(5)P-4-kinase isoform 2-beta
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human PIP5K2B (NP_003550.1).,, Sequence:, MSSNCTSTTAVAVAPLSASKTKTKKKHFVCQKVKLFRASEPILSVLMWGVNHTINELSNVPVPVMLMPDDFKAYSKIKVDNHLFNKENLPSRFKFKEYCPMVFRNLRERFGIDDQDYQNSVTRSAPINSDSQGRCGTRFLTTYDRRFVIKTVSSEDVAEMHNILKKYHQFIVECHGNTLLPQFLGMYRLTVDGVETYMVVTRNVFSHRLTVHRKYDLKGSTVAREASDKEKAKDLPTFKDNDFLNEGQKLHVGEESKKNFLEKLKRDVEF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8396
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.