missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PIPES, 1.0M buffer soln., pH 8.0

Product Code. 15463359
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463359 500 mL 500mL
15453359 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463359 Supplier Thermo Scientific Chemicals Supplier No. J60618.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES was used in the preparation of a variety of marine and mammalian tissues for electron microscopic study. Lipid loss and artifacts are minimized when PIPES was used to buffer the glutaraldehyde fixative. Fixation of fungal zoospores for fluorescence microscopy and electron microscopy was optimal with a combination of glutaraldehyde and formaldehyde in PIPES buffer. Alkaline phosphatase retention activity was 60% greater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES was used in the preparation of a variety of marine and mammalian tissues for electron microscopic study. Lipid loss and artifacts are minimized when PIPES was used to buffer the glutaraldehyde fixative. Fixation of fungal zoospores for fluorescence microscopy and electron microscopy was optimal with a combination of glutaraldehyde and formaldehyde in PIPES buffer. Alkaline phosphatase retention activity was 60% greater.

Solubility
Fully misicible in water. Soluble in water (0.05 M), giving a clear, colorless solution (the resulting solution has a pH of approximately 4.5-4.7). 10 g will dissolve in 40 mL of 0.1 N NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, reducing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
MDL Number MFCD00006159
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Color Colorless
Concentration 1.0 M
pH 8.0
Physical Form Liquid
Quantity 500 mL
Solubility Information Fully misicible in water. Soluble in water (0.05 M),giving a clear,colorless solution (the resulting solution has a pH of approximately 4.5-4.7). 10 g will dissolve in 40 mL of 0.1 N NaOH.
Chemical Name or Material PIPES buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.