accessibility menu, dialog, popup

UserName

NaOHSOONNSOOO

CAS RN 5625-37-6

IUPAC Name: sodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
Synonyms: sodium pipes pipes monosodium salt pipes monosodium monosodium pipes
Molecular Weight (g/mol): 324.34
Molecular Formula: C8H17N2NaO6S2
InChi Key: OGGAIRCLBMGXCZ-UHFFFAOYSA-M
SMILES: [Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Quantity 
  • (2)
  • (2)
  • (6)
  • (7)
  • (1)
  • (3)
  • (1)
  • (13)
  • (1)
  • (5)
  • (6)
Physical Form 
  • (4)
  • (26)
  • (4)
  • (3)
Concentration 
  • (14)
  • (10)
  • (2)
pH 
  • (2)
  • (2)
  • (6)
  • (4)
  • (4)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (6)
  • (4)
  • (29)
  • (5)
  • (2)
  • (2)
  • (6)
  • (7)
  • (1)
  • (3)
  • (1)
  • (13)
  • (1)
  • (5)
  • (6)
  • (4)
  • (26)
  • (4)
  • (3)
  • (14)
  • (10)
  • (2)
  • (2)
  • (2)
  • (6)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (3)
  • (1)
  • (4)
  • (3)