missing translation for 'onlineSavingsMsg'
Learn More

PIPES, 98%

Product Code. 11433107
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433107 25 g 25g
11443107 100 g 100g
11453107 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433107 Supplier Thermo Scientific Alfa Aesar Supplier No. A16090.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

PIPES is commonly used as a buffering agent in biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES is commonly used as a buffering agent in biochemistry.

Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
MDL Number MFCD00006159
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
Synonym pipes, 1,4-piperazinediethanesulfonic acid, piperazine-n,n'-bis 2-ethanesulfonic acid, 2,2'-piperazine-1,4-diyl diethanesulfonic acid, 1,4-piperazinebis ethanesulfonic acid, unii-g502h79v6l, 2-4-2-sulfoethyl piperazin-1-yl ethanesulfonic acid, piperazine-1,4-bis 2-ethanesulfonic acid, 1,4-piperazine-diethanesulfonic acid, 2,2'-piperazine-1,4-diyl bis ethanesulphonic acid
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 817713
Merck Index 147479
Solubility Information Soluble in 1M sodium hydroxide solution. Slightly soluble in water.
Formula Weight 302.37
Percent Purity 98%
Chemical Name or Material PIPES

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.