missing translation for 'onlineSavingsMsg'
Learn More

PIPES, 99.4%, MP Biomedicals™

Product Code. 11431741 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
500g
Product Code. Quantity unitSize
11431741 1 kg 1kg
11401741 10 g 10g
11411741 100 g 100g
11421741 500 g 500g
4 options
This item is not returnable. View return policy

Product Code. 11431741

Brand: MP Biomedicals 0219025791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
Synonym Piperazine-N, N'-bis[2-ethanesulfonic acid]
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 0.994
Formula Weight 302.4
Chemical Name or Material Pipes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.