missing translation for 'onlineSavingsMsg'
Learn More

Pirenzepine dihydrochloride, 99%, Thermo Scientific™

A M1 muscarinic receptor antagonist

40.40 € - 185.00 €

Chemical Identifiers

CAS 29868-97-1
Molecular Formula C19H23Cl2N5O2
Molecular Weight (g/mol) 424.326
MDL Number MFCD00055214
InChI Key FFNMBRCFFADNAO-UHFFFAOYSA-N
Synonym pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
PubChem CID 71405
ChEBI CHEBI:32014
IUPAC Name 11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
SMILES CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15444659
SDS View Documents
Thermo Scientific Alfa Aesar
J62252.MC
100mg
ONLINE EXCLUSIVE PRICE
41.20 €
40.40 € / 100mg
Save 0.80 €
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15424659
SDS View Documents
Thermo Scientific Alfa Aesar
J62252.03
1g
ONLINE EXCLUSIVE PRICE
189.00 €
185.00 € / 1g
Save 4.00 €
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

29868-97-1
424.326
FFNMBRCFFADNAO-UHFFFAOYSA-N
71405
11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
C19H23Cl2N5O2
MFCD00055214
pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
CHEBI:32014
CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl

Specifications

29868-97-1
MFCD00055214
14,7491
FFNMBRCFFADNAO-UHFFFAOYSA-N
11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
71405
424.32
Pirenzepine dihydrochloride
C19H23Cl2N5O2
Hygroscopic
pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl
424.326
CHEBI:32014
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 249-907-5

missing translation for 'rtecsNumber' : UU7883000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.