missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PKC gamma Polyclonal antibody specifically detects PKC gamma in Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | PKC gamma |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:100-1:500 |
| Formulation | PBS, 50% Glycerol, pH (7.3) |
| Gene Alias | EC 2.7.11, EC 2.7.11.13, MGC57564, PKCCSCA14, PKC-gamma, PKCGprotein kinase C gamma type, protein kinase C, gamma |
| Host Species | Rabbit |
| Immunogen | protein kinase C, gamma. (Uniprot# P63318) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?