missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Placental Lactogen/CSH1 Antibody (SR1403), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21927-100ul
This item is not returnable.
View return policy
Description
Placental Lactogen/CSH1 Monoclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| Placental Lactogen/CSH1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1403 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin | |
| A synthesized peptide derived from human Placental Lactogen/CSH1 (Uniprot #: P01243, P0DML2, P0DML3) | |
| 100 μg | |
| Cytokine Research, Signal Transduction | |
| 1442 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction