missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Placental Lactogen/CSH1 Antibody (SR1403), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | Placental Lactogen/CSH1 |
|---|---|
| Clone | SR1403 |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644644
|
Novus Biologicals
NBP3-21927-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645474
|
Novus Biologicals
NBP3-21927-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Placental Lactogen/CSH1 Monoclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| Placental Lactogen/CSH1 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin | |
| A synthesized peptide derived from human Placental Lactogen/CSH1 (Uniprot #: P01243, P0DML2, P0DML3) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1403 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1442 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title