missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plakophilin 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | Plakophilin 2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683652
|
Novus Biologicals
NBP2-94257-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680212
|
Novus Biologicals
NBP2-94257-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plakophilin 2 Polyclonal antibody specifically detects Plakophilin 2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Plakophilin 2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 5318 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ARVD9, MGC177501, plakophilin 2, plakophilin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 578-837 of human PKP2 (NP_001005242.2). LPEKYSQNIYIQNRNIQTDNNKSIGCFGSRSRKVKEQYQDVPMPEEKSNPKGVEWLWHSIVIRMYLSLIAKSVRNYTQEASLGALQNLTAGSGPMPTSVAQTVVQKESGLQHTRKMLHVGDPSVKKTAISLLRNLSRNLSLQNEIAKETLPDLVSIIPDTVPSTDLLIETTASACYTLNNIIQNSYQNARDLLNTGGIQKIMAISAGDAYASNKASKAASVLLYSLWAHTELHHAYKKAQFKKTDFVNSRTAKAYHSLKD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title