missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Cell Antibody (LIV3G11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50251
This item is not returnable.
View return policy
Description
Plasma Cell Monoclonal antibody specifically detects Plasma Cell in Human samples. It is validated for Immunohistochemistry
Spécification
| Plasma Cell | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:10 - 1:500 | |
| Plasma | |
| Pancreatic cancer related serum mucin | |
| 0.1 mL | |
| Immunology | |
| Human | |
| Purified |
| Immunohistochemistry | |
| LIV3G11 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu