missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Cell Antibody (LIV3G11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00 € - 351.00 €
Specifications
| Antigen | Plasma Cell |
|---|---|
| Clone | LIV3G11 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500 |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647498
|
Novus Biologicals
NBP2-50251-0.025ml |
0.025 mL |
190.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611158
|
Novus Biologicals
NBP2-50251 |
0.1 mL |
351.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plasma Cell Monoclonal antibody specifically detects Plasma Cell in Human samples. It is validated for ImmunohistochemistrySpecifications
| Plasma Cell | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS | |
| Pancreatic cancer related serum mucin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| LIV3G11 | |
| Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Plasma | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title