missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLD4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | POLD4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665621
|
Novus Biologicals
NBP2-94725-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629970
|
Novus Biologicals
NBP2-94725-0.1ml |
0.1 mL |
470.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POLD4 Polyclonal antibody specifically detects POLD4 in Human samples. It is validated for Western BlotSpecifications
| POLD4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 57804 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA polymerase delta smallest subunit p12, DNA polymerase delta subunit p12, p12, POLDSDNA polymerase delta subunit 4, polymerase (DNA-directed), delta 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-107 of human POLD4 (NP_066996.3). MGRKRLITDSYPVVKRREGPAGHSKGELAPELGEEPQPRDEEEAELELLRQFDLAWQYGPCTGITRLQRWCRAKQMGLEPPPEVWQVLKTHPGDPRFQCSLWHLYPL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title