missing translation for 'onlineSavingsMsg'
Learn More

Polyoma Virus Early Antigen Antibody (PyC) - BSA Free, Novus Biologicals™

Product Code. 18516993 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18516993 0.1 mg 0.1mg
18558310 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18516993 Supplier Novus Biologicals Supplier No. NB1002793

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Polyoma Virus Early Antigen Monoclonal antibody specifically detects Polyoma Virus Early Antigen in Virus samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polyoma Virus Early Antigen
Applications Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone PyC
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias Large T antigen, Middle T antigen, Polyoma virus large T antigen, Polyoma virus middle T antigen, Polyoma virus small T antigen, Polyomavirus early antigen, Polyomavirus large T antigen, Polyomavirus middle t antigen, Polyomavirus small T antigen, Small T antigen
Host Species Rat
Immunogen Polyoma virus-transformed Wistar rat fibroblast cellline REWA5/T1A1.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.