missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NDUFA2 Polyclonal antibody specifically detects NDUFA2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | NDUFA2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | B8, CD14, CIB8, CI-B8, complex I B8 subunit, Complex I-B8, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2 (8kD, B8), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2, NADH-ubiquinone oxidoreductase B8 subunit, NADH-ubiquinone oxidoreductase subunit CI-B8 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human NDUFA2 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?