Learn More
Thermo Scientific™ POROS™ XQ RoboColumn
RoboColumn™ chromatography products are small columns provided in 8-column strips.
245.00 € - 317.00 €
Specifications
Content And Storage | Store at 2°C to 8°C or at room temperature. |
---|---|
Flow Rate | 16 to 1000 cm/hr. |
Product Code | Brand | Column Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity | Price | Quantity & Availability | |||||
18436418
|
Thermo Scientific™
A30719 |
200 μL |
245.00 €
200µL |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18446448
|
Thermo Scientific™
A30720 |
600 μL |
317.00 €
600µL |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS™ XQ resin, a 50-micron strong anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that that enables >140 mg/mL dynamic binding capacity in the presence of up to 6.6 mS/cm NaCl, while delivering exceptional separation performance. A major improvement to the performance of traditional anion exchangers, POROS XQ resin sets the standard for strong anion exchange chromatography in both bind & elute or flow-through modes of operation.
Features of the resin:
- High dynamic binding capacity leading to reduced column size, smaller footprint, decreased water and buffer usage, and reduced cycling
- High resolution supporting improved product yield and purity, ultimately improving the cost of goods for the molecule being produced
- Robust salt tolerance allowing enhanced process design flexibility and streamlining of process flow
- Low operating back pressure and linear pressure versus flow responses driving flexible scalability
- Rigid polymeric bead with covalent surface chemistry for easier handling and packing, and superior physical and chemical stability enabling a more robust downstream purification process
RoboColumn is a registered trademark of ATOLL GmbH.
Specifications
Store at 2°C to 8°C or at room temperature. | |
16 to 1000 cm/hr. |