missing translation for 'onlineSavingsMsg'
Learn More

POU3F3 Antibody (CL15299) [Alexa Fluor« 700], Novus Biologicals Biologicals™

Product Code. 30531394 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30531394 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30531394 Supplier Novus Biologicals Supplier No. NBP344290AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POU3F3 Monoclonal antibody specifically detects POU3F3 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU3F3
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL15299
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias brain-1, Brain-specific homeobox/POU domain protein 1, Brn-1, BRN1brn-1, Oct-8, Octamer-binding protein 8, Octamer-binding transcription factor 8, OTF-8, OTF8oct-8, POU class 3 homeobox 3, POU domain class 3, transcription factor 3, POU domain, class 3, transcription factor 3
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P20264, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5455
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.