missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP1 alpha/PPP1A Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30765-25ug
This item is not returnable.
View return policy
Description
PP1 alpha/PPP1A Polyclonal antibody specifically detects PP1 alpha/PPP1A in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PP1 alpha/PPP1A | |
| Polyclonal | |
| Western Blot : 1:500-1:1000, Immunoprecipitation | |
| EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit | |
| A peptide conjugated to KLH | |
| 25 μg | |
| Phospho Specific, Protein Phosphatase | |
| 5499 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction