missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP1 alpha/PPP1A Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | PP1 alpha/PPP1A |
|---|---|
| Dilution | Western Blot : 1:500-1:1000, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30067001
|
Novus Biologicals™
NBP3-30765-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066886
|
Novus Biologicals™
NBP3-30765-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
PP1 alpha/PPP1A Polyclonal antibody specifically detects PP1 alpha/PPP1A in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western BlotSpezifikation
| PP1 alpha/PPP1A | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Sheep | |
| Phospho Specific, Protein Phosphatase | |
| Phosphate buffered saline | |
| 5499 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1:500-1:1000, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit | |
| A peptide conjugated to KLH | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts