missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP2A alpha/beta Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30778-100ug
This item is not returnable.
View return policy
Description
PP2A alpha/beta Polyclonal antibody specifically detects PP2A alpha/beta in Bovine,Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PP2A alpha/beta | |
| Polyclonal | |
| Western Blot : 1:500-1:1000 | |
| PPP2CA, PPP2CB, Replication protein C, RP-C, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit beta isoform | |
| A peptide conjugated to KLH | |
| 100 μg | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5515 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction