missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP2A alpha/beta Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | PP2A alpha/beta |
|---|---|
| Dilution | Western Blot : 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066914
|
Novus Biologicals™
NBP3-30778-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066798
|
Novus Biologicals™
NBP3-30778-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP2A alpha/beta Polyclonal antibody specifically detects PP2A alpha/beta in Bovine,Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| PP2A alpha/beta | |
| Western Blot | |
| Unconjugated | |
| Sheep | |
| Bovine, Human, Mouse, Rat | |
| PPP2CA, PPP2CB, Replication protein C, RP-C, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit beta isoform | |
| A peptide conjugated to KLH | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 5515 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title