missing translation for 'onlineSavingsMsg'
Learn More

PP2C alpha/PPM1A Antibody, PerCP, R&D Systems™

Product Code. 18604682 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18604682 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18604682 Supplier Novus Biologicals Supplier No. NBP298253PCP
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

PP2C alpha/PPM1A Polyclonal specifically detects PP2C alpha/PPM1A in Mouse samples. It is validated for ELISA/Immunoprecipitation/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Applications ELISA, Immunoprecipitation, Western Blot
Conjugate PerCP
Immunogen Produced in rabbits immunized with purified, recombinant Mouse PP2C alpha/PPM1A (Accession#: NP_032936.1; Met 1-Trp 382)
Regulatory Status RUO
Product Type Antibody
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.