missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL1 Antibody (08), Novus Biologicals™
Mouse Monoclonal Antibody
220.00 € - 326.00 €
Specifications
| Antigen | PPIL1 |
|---|---|
| Clone | 08 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000, Immunoprecipitation 4-6 μg/mg of lysate |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634267
|
Novus Biologicals
NBP3-06214-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610817
|
Novus Biologicals
NBP3-06214-50ul |
50 μg |
220.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPIL1 Monoclonal antibody specifically detects PPIL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunoprecipitationSpecifications
| PPIL1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000, Immunoprecipitation 4-6 μg/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 51645 | |
| IgG2a | |
| Protein A purified |
| 08 | |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| CYPL1cyclophilin-related gene 1, EC 5.2.1.8, hCyPX, MGC678, peptidyl-prolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 1, peptidylprolyl isomerase (cyclophilin)-like 1, PPIase, Rotamase PPIL1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PPIL1 (Uniprot#: Q9Y3C6; Met1-Gly166). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title