missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAMEF20 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05595-20ul
This item is not returnable.
View return policy
Description
PRAMEF20 Polyclonal antibody specifically detects PRAMEF20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PRAMEF20 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| PRAME family member 20, PRAME family member 20/21, PRAME family member 21, PRAMEF21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 67-274 of mouse PRAMEF20 (NP_001108549.1). GALKKRTPYLETQLQVVLEEVDKLLIQEVHPREYKLEVLDLRSVGQNYLNVWPGAMDDWLPKTTQTDPCCSKTVMTQPLKVLVDLQLNNRGQSRLFSYLVQWSDRRKGLLQLYCNKLQIWSPSQQNYRKLSQKVNLDYVETLGLHSSCSPSFLLNFAPYLRQMRNLRKLALSNIREEHIISPEKRRCIITRFTLQILKLESLQKLHLD | |
| 20 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 645425 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction