missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAMEF20 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00 € - 463.00 €
Specifications
| Antigen | PRAMEF20 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642978
|
Novus Biologicals
NBP3-05595-100ul |
100 μg |
463.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656018
|
Novus Biologicals
NBP3-05595-20ul |
20 μg |
201.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRAMEF20 Polyclonal antibody specifically detects PRAMEF20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PRAMEF20 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| PRAME family member 20, PRAME family member 20/21, PRAME family member 21, PRAMEF21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 67-274 of mouse PRAMEF20 (NP_001108549.1). GALKKRTPYLETQLQVVLEEVDKLLIQEVHPREYKLEVLDLRSVGQNYLNVWPGAMDDWLPKTTQTDPCCSKTVMTQPLKVLVDLQLNNRGQSRLFSYLVQWSDRRKGLLQLYCNKLQIWSPSQQNYRKLSQKVNLDYVETLGLHSSCSPSFLLNFAPYLRQMRNLRKLALSNIREEHIISPEKRRCIITRFTLQILKLESLQKLHLD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 645425 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title