missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pramipexole Dihydrochloride Monohydrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Pramipexole Dihydrochloride Monohydrate
Specifications
Specifications
| Chemical Name or Material | Pramipexole dihydrochloride monohydrate |
| CAS | 191217-81-9 |
| Analyte or Component Names | Pramipexole dihydrochloride monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H17 N3 S . 2 Cl H . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Pramipexole dihydrochloride monohydrate, (6S)-6-N-Propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine dihydrochloride monohydrate, 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, hydrochloride, hydrate (1:2:1), (6S)-, 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, dihydrochloride, monohydrate, (6S)- (9CI), 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, dihydrochloride, monohydrate, (S)-, Mirapex, Pramipexole dihydrochloride monohydrate, Sifrol |
| InChI Formula | InChI=1S/C10H17N3S.2ClH.H2O/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8;;;/h7,12H,2-6H2,1H3,(H2,11,13);2*1H;1H2/t7-;;;/m0.../s1 |
| SMILES | O.Cl.Cl.CCCN[C@H]1CCc2nc(N)sc2C1 |
| IUPAC Name | (6S)-6-N-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine;hydrate;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction