missing translation for 'onlineSavingsMsg'
Learn More

Pramoxine N-Oxide, TRC

Product Code. 30427199
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427199 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427199 Supplier Toronto Research Chemicals Supplier No. TRCP70085525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pramoxine N-Oxide
Analyte or Component Names Pramoxine N-Oxide
Shipping Condition Room Temperature
Molecular Formula C17 H27 N O4
Synonym Pramoxine N-Oxide
InChI Formula InChI=1S/C17H27NO4/c1-2-3-12-21-16-5-7-17(8-6-16)22-13-4-9-18(19)10-14-20-15-11-18/h5-8H,2-4,9-15H2,1H3
Solution Type Neat
SMILES CCCCOc1ccc(OCCC[N+]2([O-])CCOCC2)cc1
IUPAC Name 4-[3-(4-butoxyphenoxy)propyl]-4-oxidomorpholin-4-ium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 309.4
Quantity 25 mg
Formula Weight 309.194
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.