missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM9 Antibody (RAB-C370) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
399.00 € - 849.00 €
Specifications
| Antigen | PRDM9 |
|---|---|
| Clone | RAB-C370 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654834
|
Novus Biologicals
NBP3-20140-0.025mg |
0.025 mg |
399.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674354
|
Novus Biologicals
NBP3-20140-0.1mg |
0.1 mg |
849.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRDM9 Monoclonal antibody specifically detects PRDM9 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| PRDM9 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 56979 | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| RAB-C370 | |
| ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| Unconjugated | |
| Human | |
| Human | |
| EC 2.1.1.43, histone-lysine N-methyltransferase PRDM9, minisatellite binding protein 3 (115kD), minisatellite binding protein 3, 115kDa, PFM6MEISETZ, PR domain containing 9, PR domain zinc finger protein 9, PR domain-containing protein 9, PRMD9, ZNF899 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM9 protein under non-denaturing conditions. | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title