missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prednicarbate 11-Propanoate, Mikromol
Click to view available options
Quantity:
25 mg
Description
Prednicarbate 11-Propanoate, Mikromol
Specifications
Specifications
| Chemical Name or Material | Prednicarbate 11-Propanoate |
| Analyte or Component Names | Prednicarbate 11-Propanoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H40 O9 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3,20-Dioxo-11β-pregna-1,4-diene-11,17,21-triyl 17-(ethyl carbonate) 11,21-dipropanoate, Prednicarbate Imp. G (EP) |
| InChI Formula | InChI=1S/C30H40O9/c1-6-24(33)37-17-23(32)30(39-27(35)36-8-3)14-12-21-20-10-9-18-15-19(31)11-13-28(18,4)26(20)22(16-29(21,30)5)38-25(34)7-2/h11,13,15,20-22,26H,6-10,12,14,16-17H2,1-5H3/t20-,21-,22-,26+,28-,29-,30-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCOC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@H](C[C@]12C)OC(=O)CC)C(=O)COC(=O)CC |
| IUPAC Name | [2-[(8S,9S,10R,11S,13S,14S,17R)-17-ethoxycarbonyloxy-10,13-dimethyl-3-oxo-11-propanoyloxy-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction