missing translation for 'onlineSavingsMsg'
Learn More

Prednicarbate, Mikromol™

Product Code. 17992156
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17992156

Brand: Mikromol MM3382.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Prednicarbate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prednicarbate
CAS 73771-04-7
Analyte or Component Names Prednicarbate
Shipping Condition Room Temperature
Molecular Formula C27 H36 O8
Certifications/Compliance ISO 17034
Synonym Prednicarbate, 11beta-Hydroxy-3,20-dioxopregna-1,4-diene-17,21-diyl 17-ethylcarbonate 21-propanoate, (11beta)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Dermatop, EsCort, HOE 777, Prednisolone 17-(ethyl carbonate) 21-propionate, Prednitop, Regenit, S 77-0777
InChI Formula InChI=1S/C27H36O8/c1-5-22(31)34-15-21(30)27(35-24(32)33-6-2)12-10-19-18-8-7-16-13-17(28)9-11-25(16,3)23(18)20(29)14-26(19,27)4/h9,11,13,18-20,23,29H,5-8,10,12,14-15H2,1-4H3/t18-,19-,20-,23+,25-,26-,27-/m0/s1
SMILES CCOC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)CC
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-17-ethoxycarbonyloxy-11-hydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Molecular Weight (g/mol) 488.57
Quantity 250 mg
Formula Weight 488.241 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.