missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prednisolone Sodium Phosphate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Prednisolone Sodium Phosphate |
| CAS | 125-02-0 |
| Analyte or Component Names | Prednisolone Sodium Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 O8 P . 2 Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Prednisolone Sodium Phosphate, Ak-pred, Codelsol, Disodium prednisolone 21-phosphate, Dojilon, Hefasolon, Hydeltrasol, Inflamase, Metreton, Optival, Orapred, Pediapred, Phortisolone, Prednesol, Prednisolone 21-(disodium phosphate), Prednisolone 21-phosphate disodium, Prednisolone 21-phosphate disodium salt, Prednisolone disodium phosphate, Prednisolone sodium phosphate, Predonema, Predsol, Predsolan, Prozorin, Sodium prednisolone phosphate, Solu-Predalone, Solucort, (11beta)-11,17-Dihydroxy-21-(phosphonooxy)pregna-1,4-diene-3,20-dione disodium salt, 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-(dihydrogen phosphate) disodium salt, 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-phosphate disodium salt |
| InChI Formula | InChI=1S/C21H29O8P.2Na/c1-19-7-5-13(22)9-12(19)3-4-14-15-6-8-21(25,17(24)11-29-30(26,27)28)20(15,2)10-16(23)18(14)19;;/h5,7,9,14-16,18,23,25H,3-4,6,8,10-11H2,1-2H3,(H2,26,27,28);;/q;2*+1/p-2/t14-,15-,16-,18+,19-,20-,21-;;/m0../s1 |
| Solution Type | Neat |
| SMILES | [Na+].[Na+].C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)COP(=O)([O-])[O-] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction