missing translation for 'onlineSavingsMsg'
Learn More

Pregnanediol (5beta-Pregnane-3alpha,20(S)-diol), LoGiCal

Product Code. 30463543
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463543 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463543 Supplier LoGiCal Supplier No. LGCFOR0222.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Pregnanediol (5beta-Pregnane-3alpha,20(S)-diol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pregnanediol
CAS 80-92-2
Analyte or Component Names Pregnanediol
Shipping Condition Room Temperature
Molecular Formula C21 H36 O2
Certifications/Compliance ISO 9001
Synonym Pregnanediol, 5 beta-Pregnane-3 alpha,20-diol, 5 beta-Pregnane-3 alpha,20 alph-diol, 3 alpha,20 alpha-Dihydroxy-5 beta-pregnane, NSC 1612, NSC 47462
InChI Formula InChI=1 S/C21H36O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h13-19,22-23 H,4-12H2,1-3H3/t13-,14+,15+,16-,17+,18-,19-,20-,21+/m0/s1
SMILES C[C@H](O)[C@H]1 CC[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C
IUPAC Name (3 R,5 R,8 R,9 S,10 S,13 S,14 S,17 S)-17-[(1 S)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-3-ol
Industry Type Forensics
Molecular Weight (g/mol) 320.51
Quantity 10 mg
Formula Weight 320.2715 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.