missing translation for 'onlineSavingsMsg'
Learn More

Pregnenolone 0.1 mg/ml in Acetonitrile, LoGiCal

Product Code. 30457800
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457800 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457800 Supplier LoGiCal Supplier No. LGCAMP0222.1612
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Pregnenolone 0.1 mg/ml in Acetonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pregnenolone
CAS 145-13-1
Analyte or Component Names Pregnenolone
Shipping Condition Ice Pack (-20°C)
Molecular Formula C21 H32 O2
Certifications/Compliance ISO 17034
Synonym 5-Pregnenolone, Arthenolone, Bina-Skin, Enelone, NSC 1616, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one, Delta5-Pregnene-3 beta-ol-20-one
InChI Formula InChI=1 S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23 H,5-12H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1
SMILES CC(=O)[C@H]1 CC[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C
IUPAC Name 1-[(3 S,8 S,9 S,10 R,13 S,14 S,17 S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-17-yl]ethanone
Solvent or Matrix Acetonitrile
Industry Type Food Analysis
Molecular Weight (g/mol) 316.48
Quantity 1 mL
Formula Weight 316.2402 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.