Learn More
Invitrogen™ PRKAR2A Recombinant Rabbit Monoclonal Antibody (24GB1265)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557622
 View more versions of this product
Product Code. 30275550
  • 564.00 € / 100µL
Description
PRKAR2A Recombinant Monoclonal Antibody for Western Blot, Flow
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. It may interact with various A-kinase anchoring proteins and determine the subcellular localization of AMPK.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRKAR2A
Recombinant Monoclonal
Unconjugated
PRKAR2A
1110061A24Rik; AI317181; AI836829; cAMP-dependent protein kinase regulatory subunit RII alpha; cAMP-dependent protein kinase type II-alpha regulatory subunit; cAMP-dependent protein kinase, regulatory subunit alpha 2; MGC3606; PKR2; PRKAR2; Prkar2a; protein kinase A, RII-alpha subunit; protein kinase cAMP-dependent regulatory type II alpha; protein kinase cAMP-dependent type II regulatory subunit alpha; protein kinase, cAMP dependent regulatory, type II alpha; protein kinase, cAMP-dependent, regulatory subunit type II alpha; protein kinase, cAMP-dependent, regulatory, type 2, alpha; protein kinase, cAMP-dependent, regulatory, type II, alpha; RII(alpha); unnamed protein product
Rabbit
Affinity chromatography
RUO
5576
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1265
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P13861
PRKAR2A
A synthesized peptide derived from human PKA R2 (aa 312-404).