missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proadifen Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Proadifen Hydrochloride |
| CAS | 62-68-0 |
| Analyte or Component Names | Proadifen Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H31 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Valeric acid, 2,2-diphenyl-, 2-(diethylamino)ethyl ester hydrochloride (6CI,8CI), 2-(Diethylamino)ethyl 2,2-diphenylvalerate hydrochloride, 2'-Diethylaminoethyl 2,2-diphenylpentanoate hydrochloride, Proadifen hydrochloride, RP 5171, SKF 525, SKF 525A, U 5446, Benzeneacetic acid, alpha-phenyl-alpha-propyl-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzeneacetic acid, alpha-phenyl-alpha-propyl-, 2-(diethylamino)ethyl ester, hydrochloride (9CI), beta-Diethylaminoethyl diphenylpropylacetate hydrochloride |
| InChI Formula | InChI=1S/C23H31NO2.ClH/c1-4-17-23(20-13-9-7-10-14-20,21-15-11-8-12-16-21)22(25)26-19-18-24(5-2)6-3;/h7-16H,4-6,17-19H2,1-3H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CCCC(C(=O)OCCN(CC)CC)(c1ccccc1)c2ccccc2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction