missing translation for 'onlineSavingsMsg'
Learn More

Proadifen hydrochloride

Product Code. 15495989
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495989 1 g 1g
15415999 500 mg 500mg
15405999 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495989 Supplier Thermo Scientific Alfa Aesar Supplier No. J63833.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62-68-0 | C23H32ClNO2 | 389.964 g/mol

Proadifen hydrochloride is an inhibitor of NOS1, AChR, cytochrome P-450, and KIR6. Proadifen hydrochloride bind to the protein moiety of P-450, also reduced the hypoxic response. of Proadifen hydrochloride have good antiarrhythmic effects. SKF-525A substantially reduced the depletion of cardiac norepinephrine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Proadifen hydrochloride is an inhibitor of NOS1, AChR, cytochrome P-450, and KIR6. Proadifen hydrochloride bind to the protein moiety of P-450, also reduced the hypoxic response. of Proadifen hydrochloride have good antiarrhythmic effects. SKF-525A substantially reduced the depletion of cardiac norepinephrine.

Solubility
Soluble in methanol or water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-68-0
Molecular Formula C23H32ClNO2
Molecular Weight (g/mol) 389.964
MDL Number MFCD00055151
InChI Key FHIKZROVIDCMJA-UHFFFAOYSA-N
Synonym proadifen hydrochloride, proadifen hcl, propyladiphenin, skf-525a hydrochloride, skf 525a, sk&f 525-a, proadifen hydrochloride usan, skf 525-a, skf-525a, hydrochloride
PubChem CID 65341
IUPAC Name 2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
SMILES CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl

Specifications

Melting Point 122°C to 123°C
Color White
Quantity 1 g
Solubility Information Soluble in methanol or water
Formula Weight 389.97
Physical Form Powder
Chemical Name or Material Proadifen hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.