accessibility menu, dialog, popup

UserName

HClH3COONCH3CH3

CAS RN 62-68-0

IUPAC Name: hydrogen 2-(diethylamino)ethyl 2,2-diphenylpentanoate chloride
Synonyms: proadifen
Molecular Weight (g/mol): 389.96
Molecular Formula: C23H32ClNO2
InChi Key: FHIKZROVIDCMJA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCC(C(=O)OCCN(CC)CC)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (14)
  • (3)
Quantity 
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (3)
Physical Form 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (7)
  • (14)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)