missing translation for 'onlineSavingsMsg'
Learn More

Prochlorperazine Maleate EP Impurity A, Mikromol™

Product Code. 30461203
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461203 4 x 25 mg 100mg
30052607 25 mg 25mg
17962576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30461203 Supplier Mikromol Supplier No. MM0411.0200254AMP
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Prochlorperazine Maleate EP Impurity A, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prochlorperazine Sulfoxide
CAS 10078-27-0
Analyte or Component Names Prochlorperazine Sulfoxide
Shipping Condition Room Temperature
Molecular Formula C20 H24 Cl N3 O S
Certifications/Compliance ISO 9001
Synonym 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-10H-phenothiazine 5-oxide, 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine 5-oxide, Meterazine sulfoxide, Prochlorperazine sulfoxide, Prochlorperazine Maleate Impurity A, Prochlorperazine Impurity A
InChI Formula InChI=1S/C20H24ClN3OS/c1-22-11-13-23(14-12-22)9-4-10-24-17-5-2-3-6-19(17)26(25)20-8-7-16(21)15-18(20)24/h2-3,5-8,15H,4,9-14H2,1H3
Solution Type Neat
SMILES CN1CCN(CCCN2c3ccccc3S(=O)c4ccc(Cl)cc24)CC1
IUPAC Name 2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine 5-oxide
Molecular Weight (g/mol) 389.94
Quantity 4 x 25 mg
Formula Weight 389.1329 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.