Learn More
enQuireBio™ Recombinant Human ITLN1 / Omentin Protein
Recombinant Protein
Brand:  enQuireBio™ QP5262-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15975789
  • 7105.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.
Specifications
NP_060095.2
ITLN1 / Omentin Protein
Research Use Only
ITLN1
Human
Untagged
10 mM sodium phosphate and 5:1 mannitol to protein, pH 7.5
55600
1 mg
Less than or equal to 5.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MNQLSFLLFL IATTRGWSTD EANTYFKEWT CSSSPSLPRS CKEIKDECPS AFDGLYFLRT ENGVIYQTFC DMTSGGGGWT LVASVHENDM RGKCTVGDRW SSQQGSKADY PEGDGNWANY NTFGSAEAAT SDDYKNPGYY DIQAKDLGIW HVPNKSPMQH WRNSSLLRYR TDTGFLQTLG HNLFGIYQKY PVKYGEGKCW TDNGPVIPVV YDFGDAQKTA SYYSPYGQRE FTAGFVQFRV FNNERAANAL CAGMRVTGCN TEHHCIGGGG YFPEASPQQC GDFSGFDWSG YGTHVGYSSS REITEAAVLL FYR
Greater than 90 as determined by SDS-PAGE.