missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1QTNF9 Protein
A cDNA sequence encoding the C1QTNF9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11236-1mg
Specifications
C1QTNF9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS QDTCRQGHPG IPGNPGHNGL PGRDGRDGAK GDKGDAGEPG RPGSPGKDGT SGEKGERGAD GKVEAKGIKG DQGSRGSPGK HGPKGLAGPM GEKGLRGETG PQGQKGNKGD VGPTGPEGPR GNIGPLGPTG LPGPMGPIGK PGPKGEAGPT GPQGEPGVRG IRGWKGDRGE KGKIGETLVL PKSAFTVGLT VLSKFPSSDM PIKFDKILYN EFNHYDTAAG KFTCHIAGVY YFTYHITVFS RNVQVSLVKN GVKILHTKDA YMSSEDQASG GIVLQLKLGD EVWLQVTGGE RFNGLFADED DDTTFTGFLL FSSP |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C1QTNF9 Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 0.03M Acetate buffer, pH 4. |