missing translation for 'onlineSavingsMsg'
Learn More

Progesterone, Dr. Ehrenstorfer

Product Code. 30325290
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325290 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325290 Supplier Dr. Ehrenstorfer Supplier No. DREC16342000100MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Progesterone
CAS 57-83-0
Analyte or Component Names Progesterone
Shipping Condition +20°C
Molecular Formula C21 H30 O2
Certifications/Compliance ISO 17034
Synonym Progesterone, Pregn-4-ene-3,20-dione, Progesterone (8CI), 4-Pregnene-3,20-dione, Agolutin, Bio-luton, CIDR, Corlutin, Corlutina, Corluvite, Corporin, Corpus luteum hormone, Crinone, Cue-Mate, Cyclogest, Duraprogen, EAZI-BREED CIDR, Endometrin, Estima, Flavolutan, Fologenon, Gesterol, Gestiron, Gestone, Gestormone, Gestron, Glanducorpin, Gynlutin, Gynolutone, Hormoflaveine, Hormoluton, Lipo-Lutin, Lucorteum Sol, Lugesteron, Lugesterone, Luteal Hormone, Luteinique, Luteocrin normale, Luteodyn, Luteogan, Luteohormone, Luteol, Luteopur, Luteosan, Luteostab, Luteosterone, Luteovis, Luteum, Lutex, Lutidon, Lutin, Lutinus, Lutociclina, Lutocuclin M, Lutocyclin, Lutocyclin M, Lutocylin, Lutoform, Lutogyl, Lutren, Lutromone, Microgest, Minagest, NSC 64377, NSC 9704, Nalutron, P4, Percutacrine Luteinique, Piaponon, Primolut, Prochieve, Progeffik, Progeffik 200, Progekan, Progestan, Progestasert, Progesterol, Progestin, Progestogel, Progestol, Progeston, Progestone, Progestron, Prolets, Prolidon, Proluton, Prometrium, Prontogest, Protormone, Sincrogest, Syngesterone, Syngestrets, Syntolutan, U 3672, Utrogest, Utrogestan, Vitarrine, Delta4-Pregnene-3,20-dione
InChI Formula InChI=1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (8S,9S,10R,13S,14S,17S)-17-acetyl-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 314.46
Quantity 100 mg
Formula Weight 314.2246 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.