missing translation for 'onlineSavingsMsg'
Learn More

Progesterone, Mikromol™

Product Code. 18000410
Cambiar vista
Click to view available options
Quantity:
250 mg
Tamaño de la unidad:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
18000410 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18000410 Proveedor Mikromol N.º de proveedor MM0222.000250
 Restricted Item
Estimated Shipment: 26-11-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Progesterone
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Progesterone
CAS 57-83-0
Analyte or Component Names Progesterone
Shipping Condition Room Temperature
Molecular Formula C21 H30 O2
Certifications/Compliance ISO 17034
Synonym Progesterone, Pregn-4-ene-3,20-dione, Progesterone (8CI), 4-Pregnene-3,20-dione, Agolutin, Bio-luton, CIDR, Corlutin, Corlutina, Corluvite, Corporin, Corpus luteum hormone, Crinone, Cue-Mate, Cyclogest, Duraprogen, EAZI-BREED CIDR, Endometrin, Estima, Flavolutan, Fologenon, Gesterol, Gestiron, Gestone, Gestormone, Gestron, Glanducorpin, Gynlutin, Gynolutone, Hormoflaveine, Hormoluton, Lipo-Lutin, Lucorteum Sol, Lugesteron, Lugesterone, Luteal Hormone, Luteinique, Luteocrin normale, Luteodyn, Luteogan, Luteohormone, Luteol, Luteopur, Luteosan, Luteostab, Luteosterone, Luteovis, Luteum, Lutex, Lutidon, Lutin, Lutinus, Lutociclina, Lutocuclin M, Lutocyclin, Lutocyclin M, Lutocylin, Lutoform, Lutogyl, Lutren, Lutromone, Microgest, Minagest, NSC 64377, NSC 9704, Nalutron, P4, Percutacrine Luteinique, Piaponon, Primolut, Prochieve, Progeffik, Progeffik 200, Progekan, Progestan, Progestasert, Progesterol, Progestin, Progestogel, Progestol, Progeston, Progestone, Progestron, Prolets, Prolidon, Proluton, Prometrium, Prontogest, Protormone, Sincrogest, Syngesterone, Syngestrets, Syntolutan, U 3672, Utrogest, Utrogestan, Vitarrine, Delta4-Pregnene-3,20-dione
InChI Formula InChI=1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (8S,9S,10R,13S,14S,17S)-17-acetyl-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 314.46
Quantity 250 mg
Formula Weight 314.2246 g/mol
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.