missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Promazine Hydrochloride 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Promazine hydrochloride |
| CAS | 53-60-1 |
| Analyte or Component Names | Promazine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N2 S . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Chlorpromazine Hydrochloride Imp. C (EP), Chlorpromazine Imp. C (EP), 3-(10 H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Promazine Hydrochloride, Chlorpromazine Hydrochloride Imp. C (EP) as Hydrochloride, Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI) |
| InChI Formula | InChI=1 S/C17H20N2S.ClH/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)20-17-11-6-4-9-15(17)19;/h3-6,8-11 H,7,12-13H2,1-2H3;1 H |
| SMILES | Cl.CN(C)CCCN1c2ccccc2Sc3ccccc13 |
| IUPAC Name | N,N-dimethyl-3-phenothiazin-10-ylpropan-1-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction