missing translation for 'onlineSavingsMsg'
Learn More

Promethazine EP Impurity D, Mikromol™

Product Code. 30051678
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051678 25 mg 25mg
17990816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051678 Supplier Mikromol Supplier No. MM0412.040025
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Promethazine EP Impurity D
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Promethazine Sulfoxide
CAS 7640-51-9
Analyte or Component Names Promethazine Sulfoxide
Shipping Condition Room Temperature
Molecular Formula C17 H20 N2 O S
Certifications/Compliance ISO 17025
Synonym (2RS)-N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine S-Oxide (Promethazine Sulphoxide), (2RS)-N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine S-Oxide, Promethazine Hydrochloride Imp. D (EP), Promethazine Sulphoxide, Promethazine Hydrochloride Impurity D, Promethazine Impurity D
InChI Formula InChI=1S/C17H20N2OS/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)21(20)17-11-7-5-9-15(17)19/h4-11,13H,12H2,1-3H3
SMILES CC(CN1c2ccccc2S(=O)c3ccccc13)N(C)C
IUPAC Name N,N-dimethyl-1-(5-oxophenothiazin-10-yl)propan-2-amine
Molecular Weight (g/mol) 300.42
Quantity 25 mg
Formula Weight 300.1296 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.