missing translation for 'onlineSavingsMsg'
Learn More

Prominin 2 Antibody (244029), Biotin, R&D Systems™

Product Code. 18615423 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18615423 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18615423 Supplier Novus Biologicals Supplier No. FAB2024B
  • ONLINE EXCLUSIVE PRICE
    763.24 €
    660.45 € / 0.1mL
    Save 102.79 €
    13% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Prominin 2 Monoclonal specifically detects Prominin 2 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prominin 2
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 244029
Conjugate Biotin
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias hPROML2, MGC138714, PROM-2, prominin 2, prominin-2, Prominin-like protein 2, prominin-related protein, PROML2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Prominin 2, Ala27-Leu834, Accession # AAM10541
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150696
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.